Merck
  • Structure and Function of the Cytochrome P450 Monooxygenase, Cinnamate 4-hydroxylase (C4H1) from Sorghum bicolor.
Structure and Function of the Cytochrome P450 Monooxygenase, Cinnamate 4-hydroxylase (C4H1) from Sorghum bicolor.
Plant physiology (2020-04-26)
Bixia Zhang, Kevin M Lewis, Alejandra Abril, Dmitri R Davydov, Wilfred Vermerris, Scott E Sattler, ChulHee Kang
ABSTRACT
Cinnamate 4-hydroxylase (C4H, CYP73A) is a cytochrome P450 monooxygenase associated externally with endoplasmic reticulum of plant cells. The enzyme uses NADPH-cytochrome P450 reductase (CPR) as a donor of electrons and hydroxylates cinnamic acid to form 4-coumaric acid in phenylpropanoid metabolism. In order to better understand the structure and function of this unique class of plant P450 enzymes we have characterized the enzyme C4H1 from lignifying tissues of sorghum, encoded by Sobic.002G126600. Here we report the 1.7 Å resolution crystal structure of CYP73A33, a plant C4H enzyme. The obtained structural information along with the results of the steady-state kinetic analysis and the absorption spectroscopy titration displays a high degree of similarity of both the structural and functional features of C4H with other P450 proteins. Our data also suggest the presence of a putative allosteric substrate-binding site in a hydrophobic pocket on the enzyme surface. In addition, comparing the newly resolved structure with those of well-investigated cytochromes P450 from mammals and bacteria enabled us to identify those residues of critical functional importance, and revealed a unique sequence signature that is potentially responsible for substrate specificity and catalytic selectivity of C4H.
Product Description
Avanti
Egg PE, Avanti Research - A Croda Brand 840021P
Avanti
16:0-18:1 PA, powder
Avanti
16:0-18:1 PA, 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt), chloroform
Avanti
Egg PE, Avanti Research - A Croda Brand 840021C