Merck
  • Methylated Vnn1 at promoter regions induces asthma occurrence via the PI3K/Akt/NFκB-mediated inflammation in IUGR mice.
Methylated Vnn1 at promoter regions induces asthma occurrence via the PI3K/Akt/NFκB-mediated inflammation in IUGR mice.
Biology open (2020-03-07)
Yan Xing, Hongling Wei, Xiumei Xiao, Zekun Chen, Hui Liu, Xiaomei Tong, Wei Zhou
ABSTRACT
Infants with intrauterine growth retardation (IUGR) have a high risk of developing bronchial asthma in childhood, but the underlying mechanisms remain unclear. This study aimed to disclose the role of vascular non-inflammatory molecule 1 (vannin-1, encoded by the Vnn1 gene) and its downstream signaling in IUGR asthmatic mice induced by ovalbumin. Significant histological alterations and an increase of vannin-1 expression were revealed in IUGR asthmatic mice, accompanied by elevated methylation of Vnn1 promoter regions. In IUGR asthmatic mice, we also found (i) a direct binding of HNF4α and PGC1α to Vnn1 promoter by ChIP assay; (ii) a direct interaction of HNF4α with PGC1α; (iii) upregulation of phospho-PI3K p85/p55 and phospho-AktSer473 and downregulation of phospho-PTENTyr366, and (iv) an increase in nuclear NFκB p65 and a decrease in cytosolic IκB-α. In primary cultured bronchial epithelial cells derived from the IUGR asthmatic mice, knockdown of Vnn1 prevented upregulation of phospho-AktSer473 and an increase of reactive oxygen species (ROS) and TGF-β production. Taken together, we demonstrate that elevated vannin-1 activates the PI3K/Akt/NFκB signaling pathway, leading to ROS and inflammation reactions responsible for asthma occurrence in IUGR individuals. We also disclose that interaction of PGC1α and HNF4α promotes methylation of Vnn1 promoter regions and then upregulates vannin-1 expression.
Sigma-Aldrich
GenElute Gel Extraction Kit, sufficient for 70 purifications
Sigma-Aldrich
MISSION® shRNA, Lentiviral Particles
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
SAFC
Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12, liquid, suitable for cell culture
Sigma-Aldrich
Mouse IgE ELISA Kit
Sigma-Aldrich
Mouse IL-1 β ELISA Kit, for cell and tissue lysates
Roche
First Strand cDNA Synthesis Kit for RT-PCR (AMV), sufficient for 30 reactions (including 5 control reactions), kit of 1 (10 components), suitable for RT-PCR, hotstart: no, dNTPs included
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
NuCLEAR Extraction Kit, For mammalian tissue or cultured cells