Merck
  • The ACSL3-LPIAT1 signaling drives prostaglandin synthesis in non-small cell lung cancer.
The ACSL3-LPIAT1 signaling drives prostaglandin synthesis in non-small cell lung cancer.
Oncogene (2020-02-09)
Maria Saliakoura, Inés Reynoso-Moreno, Chiara Pozzato, Matteo Rossi Sebastiano, Mirco Galié, Jürg Gertsch, Georgia Konstantinidou
ABSTRACT
Enhanced prostaglandin production promotes the development and progression of cancer. Prostaglandins are generated from arachidonic acid (AA) by the action of cyclooxygenase (COX) isoenzymes. However, how cancer cells are able to maintain an elevated supply of AA for prostaglandin production remains unclear. Here, by using lung cancer cell lines and clinically relevant KrasG12D-driven mouse models, we show that the long-chain acyl-CoA synthetase (ACSL3) channels AA into phosphatidylinositols to provide the lysophosphatidylinositol-acyltransferase 1 (LPIAT1) with a pool of AA to sustain high prostaglandin synthesis. LPIAT1 knockdown suppresses proliferation and anchorage-independent growth of lung cancer cell lines, and hinders in vivo tumorigenesis. In primary human lung tumors, the expression of LPIAT1 is elevated compared with healthy tissue, and predicts poor patient survival. This study uncovers the ACSL3-LPIAT1 axis as a requirement for the sustained prostaglandin synthesis in lung cancer with potential therapeutic value.
Product Description
Roche
KAPA2G Fast HotStart Genotyping Mix, with dye
Avanti
17:0-20:4 PI(4,5)P2, Avanti Research - A Croda Brand LM1904, powder
Avanti
17:0-20:4 PI(4)P, Avanti Research - A Croda Brand LM1901, powder
Avanti
17:0-20:4 PI(3,4)P2, Avanti Research - A Croda Brand LM1903, powder
Avanti
18:0-20:4 DG, 1-stearoyl-2-arachidonoyl-sn-glycerol, chloroform
Roche
KAPA HotStart Mouse Genotyping Kit
Avanti
17:0-20:4 PI(3,4,5)P3, Avanti Research - A Croda Brand LM1906, powder