Merck
  • Integrin αvβ1 Modulation Affects Subtype B Avian Metapneumovirus Fusion Protein-mediated Cell-Cell Fusion and Virus Infection.
Integrin αvβ1 Modulation Affects Subtype B Avian Metapneumovirus Fusion Protein-mediated Cell-Cell Fusion and Virus Infection.
The Journal of biological chemistry (2016-05-27)
Bing-Ling Yun, Xiao-Lu Guan, Yong-Zhen Liu, Yao Zhang, Yong-Qiang Wang, Xiao-Le Qi, Hong-Yu Cui, Chang-Jun Liu, Yan-Ping Zhang, Hong-Lei Gao, Li Gao, Kai Li, Yu-Long Gao, Xiao-Mei Wang
ABSTRACT
Avian metapneumovirus (aMPV) fusion (F) protein mediates virus-cell membrane fusion to initiate viral infection, which requires F protein binding to its receptor(s) on the host cell surface. However, the receptor(s) for aMPV F protein is still not identified. All known subtype B aMPV (aMPV/B) F proteins contain a conserved Arg-Asp-Asp (RDD) motif, suggesting that the aMPV/B F protein may mediate membrane fusion via the binding of RDD to integrin. When blocked with integrin-specific peptides, aMPV/B F protein fusogenicity and viral replication were significantly reduced. Specifically we identified integrin αv and/or β1-mediated F protein fusogenicity and viral replication using antibody blocking, small interfering RNAs (siRNAs) knockdown, and overexpression. Additionally, overexpression of integrin αv and β1 in aMPV/B non-permissive cells conferred aMPV/B F protein binding and aMPV/B infection. When RDD was altered to RAE (Arg-Ala-Glu), aMPV/B F protein binding and fusogenic activity were profoundly impaired. These results suggest that integrin αvβ1 is a functional receptor for aMPV/B F protein-mediated membrane fusion and virus infection, which will provide new insights on the fusogenic mechanism and pathogenesis of aMPV.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P5D2, clone P5D2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αV Antibody, extracellular, clone AV1, azide free, culture supernatant, clone AV1, Chemicon®
Sigma-Aldrich
Anti-Na+/K+ ATPase β-1 Antibody, clone C464.8, clone C464.8, Upstate®, from mouse