Merck
  • Aerobic interval training attenuates remodelling and mitochondrial dysfunction in the post-infarction failing rat heart.
Aerobic interval training attenuates remodelling and mitochondrial dysfunction in the post-infarction failing rat heart.
Cardiovascular research (2013-04-05)
Jasenka Kraljevic, Jasna Marinovic, Danijel Pravdic, Petra Zubin, Zeljko Dujic, Ulrik Wisloff, Marko Ljubkovic
ABSTRACT
Following a large myocardial infarction (MI), remaining viable muscle often undergoes pathological remodelling and progresses towards chronic heart failure. Mitochondria may also be affected by this process and, due to their functional importance, likely contribute to the progression of the disease. Aerobic interval training (AIT) has been shown effective in diminishing pathological myocardial transformation, but the effects of AIT on mitochondrial function in hearts undergoing remodelling are not known. Adult female Sprague-Dawley rats were randomized to either 8 weeks of aerobic interval treadmill running (5 days/week), which started 4 weeks after left coronary artery ligation (MI-Trained), or a sedentary group (MI-Sedentary). Echocardiography was performed before and after the 8-week period, at which point the left ventricles (LVs) were also harvested. Twelve weeks after surgery, MI-Sedentary rats had significantly lower LV fractional shortening compared with MI-Trained rats. Complex I-dependent respiration assessed in isolated LV mitochondria was decreased by ∼37% in MI-Sedentary and 17% in MI-Trained animals (group differences P < 0.05), compared with sham-operated animals. This was paralleled with diminished ATP production and increased degree of protein oxidation in MI-Sedentary rats. The enzymatic activity of complex I was also decreased to a greater extent in MI-Sedentary than in MI-Trained animals, with no evidence of its reduced expression. When complex II substrate was used, no differences among the three groups were observed. Exercise reduces LV contractile deterioration in post-infarction heart failure and alleviates the extent of mitochondrial dysfunction, which is paralleled with preserved complex I activity.
Sigma-Aldrich
Citrate Synthase Assay Kit, 1 kit sufficient for 100 reactions (using a 1 ml cuvette), 1 kit sufficient for 480 reactions (using 96 multiwell plates)
Sigma-Aldrich
Anti-Atrial Natriuretic Polypetide Antibody, alpha, from rabbit
Sigma-Aldrich
Anti-PGC-1α Mouse mAb (4C1.3), liquid, clone 4C1.3, Calbiochem®
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.