Merck
  • NHDL, a recombinant VL/VH hybrid antibody control for IgG2/4 antibodies.
NHDL, a recombinant VL/VH hybrid antibody control for IgG2/4 antibodies.
mAbs (2019-11-02)
Corinna Lau, Martin Berner McAdam, Grethe Bergseth, Algirdas Grevys, Jack Ansgar Bruun, Judith Krey Ludviksen, Hilde Fure, Terje Espevik, Anders Moen, Jan Terje Andersen, Tom Eirik Mollnes
ABSTRACT
The mechanism of action of recombinant IgG2/4 antibodies involves blocking of their target without the induction of effector functions. Examples are eculizumab (Soliris®), which is used clinically to block complement factor C5, as well as anti-human CD14 (r18D11) and anti-porcine CD14 (rMIL2) produced in our laboratory. So far, no proper IgG2/4 control antibody has been available for controlled validation of IgG2/4 antibody functions. Here, we describe the design of a recombinant control antibody (NHDL), which was generated by combining the variable light (VL) and heavy (VH) chains from two unrelated specificities. NHDL was readily expressed and purified as a stable IgG2/4 antibody, and showed no detectable specificity toward any putative antigen present in human or porcine blood. The approach of artificial VL/VH combination may be adopted for the design of other recombinant control antibodies.
Product Description
Supelco
SYPRO® Orange Protein Gel Stain
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, tablet, 10 mg substrate per tablet
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
GS System 100mM L-Methionine sulfoximine, working range 25µM to 1mM, The GS System (offered by & a registered trademark of Celltech Biologicals, Inc.) utilizes specific vectors for cell culture transfection.
Sigma-Aldrich
cis-11-Methyl-2-dodecenoic acid, ≥90.0% (HPLC)
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, With MgCl2 and CaCl2, liquid, sterile-filtered, suitable for cell culture