Merck
  • The peroxisomal AAA ATPase complex prevents pexophagy and development of peroxisome biogenesis disorders.
The peroxisomal AAA ATPase complex prevents pexophagy and development of peroxisome biogenesis disorders.
Autophagy (2017-05-20)
Kelsey B Law, Dana Bronte-Tinkew, Erminia Di Pietro, Ann Snowden, Richard O Jones, Ann Moser, John H Brumell, Nancy Braverman, Peter K Kim
ABSTRACT
Peroxisome biogenesis disorders (PBDs) are metabolic disorders caused by the loss of peroxisomes. The majority of PBDs result from mutation in one of 3 genes that encode for the peroxisomal AAA ATPase complex (AAA-complex) required for cycling PEX5 for peroxisomal matrix protein import. Mutations in these genes are thought to result in a defect in peroxisome assembly by preventing the import of matrix proteins. However, we show here that loss of the AAA-complex does not prevent matrix protein import, but instead causes an upregulation of peroxisome degradation by macroautophagy, or pexophagy. The loss of AAA-complex function in cells results in the accumulation of ubiquitinated PEX5 on the peroxisomal membrane that signals pexophagy. Inhibiting autophagy by genetic or pharmacological approaches rescues peroxisome number, protein import and function. Our findings suggest that the peroxisomal AAA-complex is required for peroxisome quality control, whereas its absence results in the selective degradation of the peroxisome. Thus the loss of peroxisomes in PBD patients with mutations in their peroxisomal AAA-complex is a result of increased pexophagy. Our study also provides a framework for the development of novel therapeutic treatments for PBDs.
Sigma-Aldrich
Hydroxychloroquine sulfate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Complement C3 Antibody, Chemicon®, from chicken
Avanti
C16 Lyso PAF, Avanti Research - A Croda Brand 878119P, powder
Avanti
24:0 Lyso PC, 1-lignoceroyl-2-hydroxy-sn-glycero-3-phosphocholine, powder
Avanti
26:0 Lyso PC, 1-hexacosanoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder
Avanti
20:0 Lyso PC, 1-arachidoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder
Avanti
22:0 Lyso PC, 1-behenoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform
Avanti
22:0 Lyso PC, Avanti Research - A Croda Brand