Merck
Engineering alginate as bioink for bioprinting.
Acta biomaterialia (2014-07-08)
Jia Jia, Dylan J Richards, Samuel Pollard, Yu Tan, Joshua Rodriguez, Richard P Visconti, Thomas C Trusk, Michael J Yost, Hai Yao, Roger R Markwald, Ying Mei
ABSTRACT
Recent advances in three-dimensional (3-D) printing offer an excellent opportunity to address critical challenges faced by current tissue engineering approaches. Alginate hydrogels have been used extensively as bioinks for 3-D bioprinting. However, most previous research has focused on native alginates with limited degradation. The application of oxidized alginates with controlled degradation in bioprinting has not been explored. Here, a collection of 30 different alginate hydrogels with varied oxidation percentages and concentrations was prepared to develop a bioink platform that can be applied to a multitude of tissue engineering applications. The authors systematically investigated the effects of two key material properties (i.e. viscosity and density) of alginate solutions on their printabilities to identify a suitable range of material properties of alginates to be applied to bioprinting. Further, four alginate solutions with varied biodegradability were printed with human adipose-derived stem cells (hADSCs) into lattice-structured, cell-laden hydrogels with high accuracy. Notably, these alginate-based bioinks were shown to be capable of modulating proliferation and spreading of hADSCs without affecting the structure integrity of the lattice structures (except the highly degradable one) after 8days in culture. This research lays a foundation for the development of alginate-based bioink for tissue-specific tissue engineering applications.
Sigma-Aldrich
Methacrylated Alginate, Medium Viscosity, Low endotoxin
Sigma-Aldrich
Low endotoxin alginate solution, Medium viscosity, 0.2 μm, sterile-filtered
Sigma-Aldrich
Low endotoxin alginate, Medium viscosity
Sigma-Aldrich
Alginate methacrylate, medium viscosity, average degree of methacrylation 25%