Merck
  • Methyl-specific isotopic labeling: a molecular tool box for solution NMR studies of large proteins.
Methyl-specific isotopic labeling: a molecular tool box for solution NMR studies of large proteins.
Current opinion in structural biology (2015-04-17)
Rime Kerfah, Michael J Plevin, Remy Sounier, Pierre Gans, Jerome Boisbouvier
ABSTRACT
Nuclear magnetic resonance (NMR) spectroscopy is a uniquely powerful tool for studying the structure, dynamics and interactions of biomolecules at atomic resolution. In the past 15 years, the development of new isotopic labeling strategies has opened the possibility of exploiting NMR spectroscopy in the study of supra-molecular complexes with molecular weights of up to 1MDa. At the core of these isotopic labeling developments is the specific introduction of [(1)H,(13)C]-labeled methyl probes into perdeuterated proteins. Here, we describe the evolution of these approaches and discuss their impact on structural and biological studies. The relevant protocols are succinctly reviewed for single and combinatorial isotopic-labeling of methyl-containing residues, and examples of applications on challenging biological systems, including high molecular weight and membrane proteins, are presented.
Sigma-Aldrich
DLAM-LVproR-13CH3 Methyl Labeling Kit
Sigma-Aldrich
DLAM-Iδ1VproS-13CH3 Methyl Labeling Kit
Sigma-Aldrich
SLAM-VproS-13CH3 Methyl Labeling Kit
Sigma-Aldrich
DLAM-LVproS-13CH3 Methyl Labeling Kit
Sigma-Aldrich
SLAM-VproS-13CHD2 Methyl Labeling Kit
Sigma-Aldrich
PLAM-AβIδ1LVProSTγ-13CH3 Methyl Labeling Kit
Sigma-Aldrich
DLAM-LVproS-13CHD2 Methyl Labeling Kit
Sigma-Aldrich
TLAM-δI1LVproS-13CH3 Methyl Labeling Kit
Sigma-Aldrich
TLAM-Iδ1LVproR-U-13C Methyl Labeling Kit
Sigma-Aldrich
QLAM-AβIδ1LVproS-13CH3 Methyl Labeling Kit
Sigma-Aldrich
SLAM-VproR-13CH3 Methyl Labeling Kit