Merck
  • Mesoporous Palladium N,N'-Bis(3-Allylsalicylidene)o-Phenylenediamine-Methyl Acrylate Resins as Heterogeneous Catalysts for the Heck Coupling Reaction.
Mesoporous Palladium N,N'-Bis(3-Allylsalicylidene)o-Phenylenediamine-Methyl Acrylate Resins as Heterogeneous Catalysts for the Heck Coupling Reaction.
Materials (Basel, Switzerland) (2019-08-21)
Claudio Mella, Cecilia C Torres, Gina Pecchi, Cristian H Campos
ABSTRACT
Palladium N,N'-bis(3-allylsalicylidene)o-phenylenediamine complex (PdAS) immobilized onto mesoporous polymeric methyl acrylate (MA) based resins (PdAS(x)-MA, x = 1, 2, 5, or 10 wt.%) were successfully prepared as heterogeneous catalysts for the Heck reaction. The catalysts were synthesized via radical suspension polymerization using PdAS as a metal chelate monomer, divinylbenzene and MA as co-monomers. The effect of the PdAS(x) content on the physicochemical properties of the resins is also reported. The catalysts were characterized by using a range of analytical techniques. The large surface area (>580 m2·g-1) and thermal stability (up to 250 °C) of the PdAS(x)-MA materials allows their application as catalysts in the C-C coupling reaction between iodobenzene and MA in the presence of trimethylamine at 120 °C using DMF as the solvent. The PdAS(10)-MA catalyst exhibited the highest catalytic performance with no significant catalytic loss being observed after five reuses, thereby indicating excellent catalyst stability in the reaction medium.
Sigma-Aldrich
3-Allylsalicylaldehyde, 97%
Sigma-Aldrich
Allyl methyl sulfone, 96%
Sigma-Aldrich
1,3-Di-o-tolylguanidine, 99%