Merck
  • Autophagy-Mediated Secretory Pathway is Responsible for Both Normal and Pathological Tau in Neurons.
Autophagy-Mediated Secretory Pathway is Responsible for Both Normal and Pathological Tau in Neurons.
Journal of Alzheimer's disease : JAD (2019-07-01)
Seokjo Kang, Sung Min Son, Sung Hoon Baik, Jinhee Yang, Inhee Mook-Jung
ABSTRACT
Increased levels of total tau (t-tau) and hyperphosphorylated tau (p-tau) proteins in the cerebrospinal fluid of Alzheimer's disease (AD) patients are well documented and strongly correlate with AD pathology. Recent studies have further shown that human tau can be released into the extracellular space and transferred to nascent neurons. However, because the tau protein has no signal peptide identity, the mechanisms underlying its secretion remain poorly understood. In the present study, we confirmed that tau protein secretion was promoted by autophagy inducers and downregulated by beclin1 knockdown or autophagy inhibitors derived from human wild type tau (wt-tau)-overexpressing SH-SY5Y cells. Moreover, both t-tau and p-tau secretion were increased by autophagy activation. Furthermore, we identified that six isoforms of tau protein are secreted in an autophagy-dependent manner. These results indicate that both normal and pathological tau are secreted via an autophagy-mediated secretory pathway in neurons. Understanding this new pathway for tau secretion may provide critical future insights into tau pathologies, such as AD.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥99% (HPLC), from Saccharomyces cerevisiae
Sigma-Aldrich
Spautin-1, ≥98% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Triton X-100
Sigma-Aldrich
Nigericin sodium salt, ≥98% (TLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human BECN1