Merck
  • Body size-dependent energy storage causes Kleiber's law scaling of the metabolic rate in planarians.
Body size-dependent energy storage causes Kleiber's law scaling of the metabolic rate in planarians.
eLife (2019-01-05)
Albert Thommen, Steffen Werner, Olga Frank, Jenny Philipp, Oskar Knittelfelder, Yihui Quek, Karim Fahmy, Andrej Shevchenko, Benjamin M Friedrich, Frank Jülicher, Jochen C Rink
ABSTRACT
Kleiber's law, or the 3/4 -power law scaling of the metabolic rate with body mass, is considered one of the few quantitative laws in biology, yet its physiological basis remains unknown. Here, we report Kleiber's law scaling in the planarian Schmidtea mediterranea. Its reversible and life history-independent changes in adult body mass over 3 orders of magnitude reveal that Kleiber's law does not emerge from the size-dependent decrease in cellular metabolic rate, but from a size-dependent increase in mass per cell. Through a combination of experiment and theoretical analysis of the organismal energy balance, we further show that the mass allometry is caused by body size dependent energy storage. Our results reveal the physiological origins of Kleiber's law in planarians and have general implications for understanding a fundamental scaling law in biology.
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
Dioleoylglycerol, ≥99%, mixture of 1,3- and 1,2-isomers, liquid
Sigma-Aldrich
Cholesteryl linoleate, ≥98% (HPLC; detection at 205 nm)
Sigma-Aldrich
Glyceryl trioleate, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, ammonium sulfate suspension, ≥40 units/mg protein
Avanti
C12 Lactosyl(β) Ceramide (d18:1/12:0), Avanti Research - A Croda Brand 860545P, powder