Merck
  • The antiemetic profile of Y-25130, a new selective 5-HT3 receptor antagonist.
The antiemetic profile of Y-25130, a new selective 5-HT3 receptor antagonist.
European journal of pharmacology (1991-04-24)
T Fukuda, M Setoguchi, K Inaba, H Shoji, T Tahara
ABSTRACT
Y-25130( (+/-)N-(1-azabicyclo[2.2.2]oct-3-yl)-6-chloro-4-methyl-3-oxo-3,4-dihydro - 2H-1,4-benzoxazine-8-carboxamide hydrochloride) is a potent and selective 5-HT3 receptor antagonist free of dopamine receptor blocking activity. This compound was effective against emesis induced in animals by cytotoxic drugs or by total body X-radiation. When given prophylactically, the doses required to completely inhibit cisplatin-induced emesis in dogs and doxorubicin and cyclophosphamide-induced emesis in ferrets were 0.1 and 0.3 mg/kg i.v., respectively. Y-25130, at the dose of 0.3 mg/kg i.v., almost completely inhibited X-radiation-induced emesis in ferrets. When given during emesis, the doses required to completely inhibit cisplatin-induced emesis in dogs and doxorubicin- and cyclophosphamide-induced emesis in ferrets were 0.1 and 0.3 mg/kg i.v., respectively. The i.v. dose of 0.3 mg/kg of Y-25130 was enough to almost completely inhibit cisplatin-induced emesis in dogs for 24 h. From these results, it is suggested that Y-25130 may become an effective antiemetic drug against emesis induced by anticancer therapy.
Sigma-Aldrich
Azasetron hydrochloride, ≥98% (HPLC)