Merck
  • Biphasic activation of p21ras by endothelin-1 sequentially activates the ERK cascade and phosphatidylinositol 3-kinase.
Biphasic activation of p21ras by endothelin-1 sequentially activates the ERK cascade and phosphatidylinositol 3-kinase.
The EMBO journal (1997-11-14)
M Foschi, S Chari, M J Dunn, A Sorokin
ABSTRACT
Endothelin-1 (ET-1) induces cell proliferation and differentiation through multiple G-protein-linked signaling systems, including p21ras activation. Whereas p21ras activation and desensitization by receptor tyrosine kinases have been extensively investigated, the kinetics of p21ras activation induced by engagement of G-protein-coupled receptors remains to be fully elucidated. In the present study we show that ET-1 induces a biphasic activation of p21ras in rat glomerular mesangial cells. The first peak of activation of p21ras, at 2-5 min, is mediated by immediate association of phosphorylated Shc with the guanosine exchange factor Sos1 via the adaptor protein Grb2. This initial activation of p21ras results in activation of the extracellular signal-regulated kinase (ERK) cascade. We demonstrate that ET-1 signaling elicits a negative feedback mechanism, modulating p21ras activity through ERK-dependent Sos1 phosphorylation, findings which were confirmed using an adenovirus MEK construct. Subsequent to p21ras and ERK deactivation, Sos1 reverts to the non-phosphorylated condition, enabling it to bind again to the Grb2/Shc complex, which is stabilized by persistent Shc phosphorylation. However, the resulting secondary activation of p21ras at 30 min does not lead to ERK activation, correlating with intensive, ET-1-induced expression of MAP kinase phosphatase-1, but does result in increased p21ras-associated phosphatidylinositol 3-kinase activity. Our data provide evidence that ET-1-induced biphasic p21ras activation causes sequential stimulation of divergent downstream signaling pathways.
Sigma-Aldrich
Ras Activation Assay Kit
Sigma-Aldrich
Ras Assay Reagent (Raf-1 RBD, agarose)
Sigma-Aldrich
Raf-1 RBD Protein, GST, 300 µg, GST fusion-protein, corresponding to the human Ras Binding Domain (RBD, residues 1-149) of Raf-1, expressed in E. coli. with purity 50% at full length molecular weight 42 kDa. For use in Affinity Binding Assays
Sigma-Aldrich
Ral Activation Assay Kit, Non-radioactive Ral Activation Assay Kit can be used to precipitate Ral-A-GTP from cell lysates & detection by a Ral-A specific antibody.