Merck
Gold nanoparticles in biology: beyond toxicity to cellular imaging.
Accounts of chemical research (2008-08-21)
Catherine J Murphy, Anand M Gole, John W Stone, Patrick N Sisco, Alaaldin M Alkilany, Edie C Goldsmith, Sarah C Baxter
ABSTRACT
Gold, enigmatically represented by the target-like design of its ancient alchemical symbol, has been considered a mystical material of great value for centuries. Nanoscale particles of gold now command a great deal of attention for biomedical applications. Depending on their size, shape, degree of aggregation, and local environment, gold nanoparticles can appear red, blue, or other colors. These visible colors reflect the underlying coherent oscillations of conduction-band electrons ("plasmons") upon irradiation with light of appropriate wavelengths. These plasmons underlie the intense absorption and elastic scattering of light, which in turn forms the basis for many biological sensing and imaging applications of gold nanoparticles. The brilliant elastic light-scattering properties of gold nanoparticles are sufficient to detect individual nanoparticles in a visible light microscope with approximately 10(2) nm spatial resolution. Despite the great excitement about the potential uses of gold nanoparticles for medical diagnostics, as tracers, and for other biological applications, researchers are increasingly aware that potential nanoparticle toxicity must be investigated before any in vivo applications of gold nanoparticles can move forward. In this Account, we illustrate the importance of surface chemistry and cell type for interpretation of nanoparticle cytotoxicity studies. We also describe a relatively unusual live cell application with gold nanorods. The light-scattering properties of gold nanoparticles, as imaged in dark-field optical microscopy, can be used to infer their positions in a living cell construct. Using this positional information, we can quantitatively measure the deformational mechanical fields associated with living cells as they push and pull on their local environment. The local mechanical environment experienced by cells is part of a complex feedback loop that influences cell metabolism, gene expression, and migration.
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and azide functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and biotin functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and methyl functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and protein A functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and NHS functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and maleimide functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and methyl functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and amine functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and protein A functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and maleimide functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and NHS functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and azide functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and amine functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and carboxyl functionalized, powder
Sigma-Aldrich
Gold nanorods, 10nm diameter, Cy3 and biotin functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and carboxyl functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm diameter, Cy3 and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and protein A functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and azide functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and carboxyl functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and NHS functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and biotin functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and maleimide functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and NHS functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and methyl functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and amine functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and protein A functionalized, powder