Merck
  • Androgen receptor drives hepatocellular carcinogenesis by activating enhancer of zeste homolog 2-mediated Wnt/β-catenin signaling.
Androgen receptor drives hepatocellular carcinogenesis by activating enhancer of zeste homolog 2-mediated Wnt/β-catenin signaling.
EBioMedicine (2018-08-29)
Haibin Song, Zhuo Yu, Xuehua Sun, Jun Feng, Qi Yu, Hanif Khan, Xiaojun Zhu, Lingying Huang, Man Li, Myth T S Mok, Alfred S L Cheng, Yueqiu Gao, Hai Feng
ABSTRACT
Androgen receptor (AR) plays a crucial role as a transcription factor in promoting the development of hepatocellular carcinoma (HCC) which is prone to aberrant chromatin modifications. However, the regulatory effects of AR on epigenetic mediators in HCC remain ill-defined. Enhancer of zeste homolog 2 (EZH2), an oncogene responsible for the tri-methylation of histone H3 at lysine 27 (H3K27me3), was identified to be overexpressed in approximate 70-90% of HCC cases, which prompted us to investigate whether or how AR regulates EZH2 expression. Colony formation, soft agar assay, xenograft and orthotopic mouse models were used to determine cell proliferation and tumorigenicity of gene-manipulated HCC cells. Gene regulation was assessed by chromatin immunoprecipitation, luciferase reporter assay, quantitative RT-PCR and immunoblotting. Clinical relevance of candidate proteins in patient specimens was examined in terms of pathological parameters and postsurgical survival rates. In this study, we found that AR upregulated EZH2 expression by binding to EZH2 promoter and stimulating its transcriptional activity. EZH2 overexpression increased H3K27me3 levels and thereby silenced the expression of Wnt signal inhibitors, resulting in activation of Wnt/β-catenin signaling and subsequently induction of cell proliferation and tumorigenesis. In a cohort of human HCC patients, concordant overexpression of AR, EZH2, H3K27me3 and active β-catenin was observed in tumor tissues compared with paired non-tumor tissues, which correlated with tumor progression and poor prognosis. These findings demonstrate a novel working model in which EZH2 mediates AR-induced Wnt/β-catenin signaling activation through epigenetic modification, and support the application of EZH2-targeted reagents for treating HCC patients.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-NKD1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-PPP2R2B antibody produced in mouse, clone 1F3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
Anti-Androgen Receptor Antibody, from rabbit, purified by affinity chromatography