Merck
Mast Cells in the Developing Brain Determine Adult Sexual Behavior.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2018-08-11)
Kathryn M Lenz, Lindsay A Pickett, Christopher L Wright, Katherine T Davis, Aarohi Joshi, Margaret M McCarthy
ABSTRACT
Many sex differences in brain and behavior are programmed during development by gonadal hormones, but the cellular mechanisms are incompletely understood. We found that immune-system-derived mast cells are a primary target for the masculinizing hormone estradiol and that mast cells are in turn primary mediators of brain sexual differentiation. Newborn male rats had greater numbers and more activated mast cells in the preoptic area (POA), a brain region essential for male copulatory behavior, than female littermates during the critical period for sexual differentiation. Inhibiting mast cells with a stabilizing agent blunted the masculinization of both POA neuronal and microglial morphology and adult sex behavior, whereas activating mast cells in females, even though fewer in number, induced masculinization. Treatment of newborn females with a masculinizing dose of estradiol increased mast cell number and induced mast cells to release histamine, which then stimulated microglia to release prostaglandins and thereby induced male-typical synaptic patterning. These findings identify a novel non-neuronal origin of brain sex differences and resulting motivated behaviors.SIGNIFICANCE STATEMENT We found that immune-system-derived mast cells are a primary target for the masculinizing hormone estradiol and that mast cells are in turn primary mediators of brain sexual differentiation. These findings identify a novel non-neuronal origin of brain sex differences and resulting motivated behaviors.
Sigma-Aldrich
Anti-MAP2 (2a+2b) antibody, Mouse monoclonal, clone AP-20, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
β-Estradiol-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Estrogen Receptor α Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Anti-Histamine Receptor 1 Antibody, CT, Chemicon®, from rabbit
Supelco
1,25-Dihydroxyvitamin D3-13C3 (25,26,27-13C3) solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®