Merck
  • The mitochondrial transporter SLC25A25 links ciliary TRPP2 signaling and cellular metabolism.
The mitochondrial transporter SLC25A25 links ciliary TRPP2 signaling and cellular metabolism.
PLoS biology (2018-08-07)
Alexis Hofherr, Claudia Seger, Fiona Fitzpatrick, Tilman Busch, Elisabeth Michel, Jingting Luan, Lea Osterried, Frieder Linden, Albrecht Kramer-Zucker, Barbara Wakimoto, Conny Schütze, Nils Wiedemann, Anna Artati, Jerzy Adamski, Gerd Walz, Edmund R S Kunji, Craig Montell, Terry Watnick, Michael Köttgen
ABSTRACT
Cilia are organelles specialized in movement and signal transduction. The ciliary transient receptor potential ion channel polycystin-2 (TRPP2) controls elementary cilia-mediated physiological functions ranging from male fertility and kidney development to left-right patterning. However, the molecular components translating TRPP2 channel-mediated Ca2+ signals into respective physiological functions are unknown. Here, we show that the Ca2+-regulated mitochondrial ATP-Mg/Pi solute carrier 25 A 25 (SLC25A25) acts downstream of TRPP2 in an evolutionarily conserved metabolic signaling pathway. We identify SLC25A25 as an essential component in this cilia-dependent pathway using a genome-wide forward genetic screen in Drosophila melanogaster, followed by a targeted analysis of SLC25A25 function in zebrafish left-right patterning. Our data suggest that TRPP2 ion channels regulate mitochondrial SLC25A25 transporters via Ca2+ establishing an evolutionarily conserved molecular link between ciliary signaling and mitochondrial metabolism.
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
Sigma-Aldrich
Pentaethylene glycol, 98%
Sigma-Aldrich
2,2-Dimethyl-1,3-propanediol, 99%