Merck
Astrocytes promote myelination in response to electrical impulses.
Neuron (2006-03-18)
Tomoko Ishibashi, Kelly A Dakin, Beth Stevens, Philip R Lee, Serguei V Kozlov, Colin L Stewart, R Douglas Fields
ABSTRACT
Myelin, the insulating layers of membrane wrapped around axons by oligodendrocytes, is essential for normal impulse conduction. It forms during late stages of fetal development but continues into early adult life. Myelination correlates with cognitive development and can be regulated by impulse activity through unknown molecular mechanisms. Astrocytes do not form myelin, but these nonneuronal cells can promote myelination in ways that are not understood. Here, we identify a link between myelination, astrocytes, and electrical impulse activity in axons that is mediated by the cytokine leukemia inhibitory factor (LIF). These findings show that LIF is released by astrocytes in response to ATP liberated from axons firing action potentials, and LIF promotes myelination by mature oligodendrocytes. This activity-dependent mechanism promoting myelination could regulate myelination according to functional activity or environmental experience and may offer new approaches to treating demyelinating diseases.
Sigma-Aldrich
Anti-O1 Antibody, clone 59, clone 59, Chemicon®, from mouse
Sigma-Aldrich
Leukemia Inhibitory Factor from mouse, 10 µg, mouse recombinant LIF protein, expressed in E. coli, suitable for stem cell culture
Sigma-Aldrich
Leukemia Inhibitory Factor from mouse, 5 µg, mouse recombinant LIF protein, expressed in E. coli, suitable for stem cell culture