Merck
Small-Molecule PROTACS: New Approaches to Protein Degradation.
Angewandte Chemie (International ed. in English) (2016-01-13)
Momar Toure, Craig M Crews
ABSTRACT
The current inhibitor-based approach to therapeutics has inherent limitations owing to its occupancy-based model: 1) there is a need to maintain high systemic exposure to ensure sufficient in vivo inhibition, 2) high in vivo concentrations bring potential for off-target side effects, and 3) there is a need to bind to an active site, thus limiting the drug target space. As an alternative, induced protein degradation lacks these limitations. Based on an event-driven model, this approach offers a novel catalytic mechanism to irreversibly inhibit protein function by targeting protein destruction through recruitment to the cellular quality control machinery. Prior protein degrading strategies have lacked therapeutic potential. However, recent reports of small-molecule-based proteolysis-targeting chimeras (PROTACs) have demonstrated that this technology can effectively decrease the cellular levels of several protein classes.
Sigma-Aldrich
2,7-Diazaspiro[3.5]nonane-7-acetic acid, 2-[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl]-, ≥95%
Sigma-Aldrich
Pomalidomide-piperazine, ≥ 95.0%
Sigma-Aldrich
VH 032 amide-alkyl C5-acid, ≥95%
Sigma-Aldrich
Pomalidomide-PEG2-C2-azide, ≥95%
Sigma-Aldrich
Pomalidomide-methylamino-PEG1-NH2 hydrochloride
Sigma-Aldrich
(S,R,S)-AHPC-CO-PEG4-C2-acid, ≥95%
Sigma-Aldrich
Thalidomide-O-amido-C8-NH2 trifluoroacetate, ≥95.0%
Sigma-Aldrich
(S,R,S)-AHPC-alkyne-piperidine hydrochloride
Sigma-Aldrich
1-Piperazinecarboxylic acid, 4-(4-piperidinyl)-, 1,1-dimethylethyl ester, ≥98%
Sigma-Aldrich
4-(Aminoethyl)-1-N-Boc-piperidine, ≥95.0%
Sigma-Aldrich
(S,R,S)-AHPC-acetamido-O-PEG4-C1-acid, ≥95%
Sigma-Aldrich
Pomalidomide-piperidine-carboxylic acid, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-PEG6-NH2 hydrochloride, ≥95%
Sigma-Aldrich
Pomalidomide-PEG2-NH2 hydrochloride, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-C6-PEG3-butyl amine hydrochloride, ≥95%
Sigma-Aldrich
Pomalidomide-PEG6-butyl CO2H, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-PEG2-NH2 hydrochloride, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-PEG1-NH2 hydrochloride, ≥95%
Sigma-Aldrich
Pomalidomide-PEG4-NH2 hydrochloride, ≥95%
Sigma-Aldrich
Pomalidomide-C2-NH2 hydrochloride, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-C9-NH2 hydrochloride, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-pentanoic-acid, ≥95%
Sigma-Aldrich
Pomalidomide-difluoroPEG1-C4-piperazine Hydrochloride, ≥95%
Sigma-Aldrich
Propanoic acid, 3-[2-[2-[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl]amino]ethoxy]ethoxy]ethoxy]-, ≥95%
Sigma-Aldrich
C5 Lenalidomide-methylamino-PEG1-NH2 hydrochloride, ≥95%
Sigma-Aldrich
Pomalidomide-pyridine-PEG1-piperazine hydrochloride
Sigma-Aldrich
(S,R,S)-AHPC-pyrimidine-piperazine-PEG1-NH2 hydrochloride
Sigma-Aldrich
FBnG-C3-PEG5-C3-NH2 hydrochloride, ≥95%
Sigma-Aldrich
VH032-cyclopropane-F, ≥95%
Sigma-Aldrich
Pomalidomide-PEG5-NH2 hydrochloride