Merck
  • Analysis of diltiazem and desacetyldiltiazem in serum by high-performance liquid chromatography.
Analysis of diltiazem and desacetyldiltiazem in serum by high-performance liquid chromatography.
Polish journal of pharmacology (1995-09-01)
D Paczkowski, R Dabrowski, D Sitkiewicz, Z Sadowski
ABSTRACT
This paper describes a simple high-performance liquid chromatographic (HPLC) method for the determination of diltiazem and desacetyldiltiazem in human serum. After basic methyl-tert-butyl ether extraction and back-extraction with hydrochloric acid, the drug and its metabolite was injected into a Supelcosil LC-CN column and the absorbance of the eluate was measured at 240 nm. The sensitivity was 5 ng/ml and the obtained precision, selectivity and stability during storage were adequate for the performed clinical studies in patients therapeutically treated with diltiazem.
Product Description
Supelco
SUPELCOSIL LC-CN HPLC Column, 3 μm particle size, L × I.D. 3.3 cm × 4.6 mm
Supelco
SUPELCOSIL LC-CN HPLC Column, 3 μm particle size, L × I.D. 3.3 cm × 3 mm
Supelco
SUPELCOSIL LC-CN HPLC Column, 3 μm particle size, L × I.D. 7.5 cm × 4.6 mm
Supelco
SUPELCOSIL LC-CN Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-CN HPLC Column, 3 μm particle size, L × I.D. 7.5 cm × 3 mm
Supelco
SUPELCOSIL LC-CN Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-CN HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL LC-CN HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm