Merck
  • Simultaneous gas chromatographic determination of carbofuran, metalaxyl, and simazine in soils.
Simultaneous gas chromatographic determination of carbofuran, metalaxyl, and simazine in soils.
Journal - Association of Official Analytical Chemists (1989-03-01)
L W Getzin, C G Cogger, P R Bristow
ABSTRACT
A method for extraction, cleanup, and simultaneous gas chromatographic detection of carbofuran, metalaxyl, and simazine in soils has been developed. Pesticide residues were extracted from soil with acetone containing 10% 0.2M HCl-KCl buffer (pH 2.0), cleaned up with methylene chloride-carbonate buffer (pH 10.7) solvent partitioning and solid-phase extraction on disposable silica gel columns, and quantitated with gas chromatography using a Supelcowax 10 megabore capillary column and a nitrogen-selective detector. Method limits of detection were 0.02 microgram/g for the 3 pesticides in surface soils (0-30 cm depths) and 0.02, 0.02, and 0.005 microgram/g in soils below 30 cm (subsoils) for carbofuran, metalaxyl, and simazine, respectively. Recoveries for carbofuran, metalaxyl, and simazine were 92.6 +/- 5.5, 93.6 +/- 5.0, and 88.4 +/- 6.7%, respectively, when soil samples were spiked with pesticide concentrations ranging from 0.02 to 2.0 micrograms/g.
Product Description
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.53 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 2.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.10 mm, df 0.10 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 2.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.20 mm, df 0.20 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 100 m × 0.25 mm, df 0.25 μm