Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Zinc nitrate hexahydrate
Linear Formula:
Zn(NO
3
)
2
· 6H
2
O
CAS Number:
10196-18-6
Molecular Weight:
297.49
EC Number:
231-943-8
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
228737
Zinc nitrate hexahydrate,
reagent grade, 98%
Expand
View Pricing
96482
Zinc nitrate hexahydrate,
purum p.a., crystallized, ≥99.0% (KT)
Expand
View Pricing