Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Zinc bromide
Synonym(s):
Zinc dibromide, Zinc bromide
Linear Formula:
ZnBr
2
CAS Number:
7699-45-8
Molecular Weight:
225.20
EC Number:
231-718-4
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (4)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade
Anhydrous (2)
Puriss (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
230022
Zinc bromide,
99.999% trace metals basis
Expand
View Pricing
8.18631
Zinc bromide,
anhydrous for synthesis
Expand
View Pricing
451398
Zinc bromide,
AnhydroBeads
™
, −10 mesh, 99.999% trace metals basis
Expand
View Pricing
02128
Zinc bromide,
puriss., anhydrous, ≥98%
Expand
View Pricing