Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Xenon-
124
Xe
Linear Formula:
124
Xe
CAS Number:
15687-60-2
Molecular Weight:
123.91
Products
Product Category
gases (7)
stable isotopes (7)
Brand
Sigma-Aldrich (7)
Formula Weight
Physical Form
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
602094
Xenon-
124
Xe,
99.9 atom %
Expand
View Pricing
602035
Xenon-
124
Xe,
5 atom %
Expand
View Pricing
602078
Xenon-
124
Xe,
90 atom %
Expand
View Pricing
602043
Xenon-
124
Xe,
10 atom %
Expand
View Pricing
602027
Xenon-
124
Xe,
1 atom %
Expand
View Pricing
602019
Xenon-
124
Xe,
70 atom %
Expand
View Pricing
602051
Xenon-
124
Xe,
50 atom %
Expand
View Pricing