Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Williams’ Medium E
Products
Product Category
media (4)
Brand
SAFC (2)
Sigma-Aldrich (2)
Physical Form
L-Glutamine
Sodium Bicarbonate
Phenol Red
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
W1878
Williams’ Medium E,
With sodium bicarbonate, without ʟ-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Expand
View Pricing
W4128
Williams’ Medium E,
With sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture
Expand
View Pricing
W4125
Williams’ Medium E,
With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Expand
View Pricing
W0397
Williams’ Medium E,
liquid, sterile-filtered, suitable for cell culture, With sodium bicarbonate and L-glutamine, without phenol red
Expand
View Pricing