Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wheaton
®
Dropping Bottle
Products
Product Category
bottles (28)
Capacity (mL)
Color
Vial Bottle Color
Package Size
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
DWKW242835
Wheaton
®
Dropping Bottle,
white bottle, capacity 15 mL
Expand
View Pricing
DWKW242837
Wheaton
®
Dropping Bottle,
white bottle, capacity 60 mL
Expand
View Pricing
DWKW242839
Wheaton
®
Dropping Bottle,
white bottle, capacity 120 mL
Expand
View Pricing
DWKW242834
Wheaton
®
Dropping Bottle,
white bottle, capacity 10 mL
Expand
View Pricing
DWKW242829
Wheaton
®
Dropping Bottle,
natural bottle, capacity 120 mL
Expand
View Pricing
DWKW242831
Wheaton
®
Dropping Bottle,
white bottle, capacity 3 mL
Expand
View Pricing
DWKW242824
Wheaton
®
Dropping Bottle,
natural bottle, capacity 10 mL
Expand
View Pricing
DWKW242821-A
Wheaton
®
Dropping Bottle,
natural bottle, capacity 3 mL, case of 1000
Expand
View Pricing
DWKW242824-A
Wheaton
®
Dropping Bottle,
natural bottle, capacity 10 mL, case of 1000
Expand
View Pricing
DWKW242821
Wheaton
®
Dropping Bottle,
natural bottle, capacity 3 mL
Expand
View Pricing
DWKW242833
Wheaton
®
Dropping Bottle,
white bottle, capacity 7 mL
Expand
View Pricing
DWKW242836
Wheaton
®
Dropping Bottle,
white bottle, capacity 30 mL
Expand
View Pricing
DWKW242831-A
Wheaton
®
Dropping Bottle,
white bottle, capacity 3 mL, case of 1000
Expand
View Pricing
DWKW242833-A
Wheaton
®
Dropping Bottle,
white bottle, capacity 7 mL, case of 1000
Expand
View Pricing
DWKW242835-A
Wheaton
®
Dropping Bottle,
white bottle, capacity 15 mL, case of 1000
Expand
View Pricing
DWKW242832
Wheaton
®
Dropping Bottle,
white bottle, capacity 6 mL
Expand
View Pricing
DWKW242825
Wheaton
®
Dropping Bottle,
natural bottle, capacity 15 mL
Expand
View Pricing
DWKW242826
Wheaton
®
Dropping Bottle,
natural bottle, capacity 30 mL
Expand
View Pricing
DWKW242832-A
Wheaton
®
Dropping Bottle,
white bottle, capacity 6 mL, case of 1000
Expand
View Pricing
DWKW242834-A
Wheaton
®
Dropping Bottle,
white bottle, capacity, case of 1000
Expand
View Pricing
Page 1 of 2
Page 1 of 2