Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
Seed testing paper
Products
Product Category
filter discs (7)
test strips (1)
Color
Manufacturer
Diameter (mm)
Length (mm)
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
WHA10344676
Whatman
®
Seed testing paper,
3014 Pleated Strips White, 50 double pleats, W × L 110 mm × 20 mm , includes wrapping strips
Expand
View Pricing
WHA2181090
Whatman
®
Seed testing paper,
Grade 181, support filter for seed test using Copenhagen tank system, diam. 90 mm
Expand
View Pricing
WHA10345576
Whatman
®
Seed testing paper,
Grade 3236, pleated strips grey, includes wrapping strips, L × W 110 mm × 20 mm
Expand
View Pricing
WHA10342583
Whatman
®
Seed testing paper,
grade, 3645 paper, yellow, 110 × 170 mm, 100/PK
Expand
View Pricing
WHA10342581
Whatman
®
Seed testing paper,
L × W (420 mm) (594 mm), pack of 50 ea
Expand
View Pricing
WHA10344672
Whatman
®
Seed testing paper,
3014 Pleated Strips White, 25 double pleats, W × L 110 mm × 20 mm
Expand
View Pricing
WHA10342580
Whatman
®
Seed testing paper,
Grade 3644, sheets, L × W 140 mm × 200 mm
Expand
View Pricing
WHA10345572
Whatman
®
Seed testing paper,
3236 Pleated Strips Grey, 25 double pleats, W × L 110 mm × 20 mm
Expand
View Pricing