Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
indicator papers
Products
Product Category
test strips (28)
Manufacturer
Length (m)
Detection Method
Package Size
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
WHA10362000
Whatman
®
indicator papers,
Indicator paper, pH range 0-14, non-bleeding
Expand
View Pricing
WHA2613991
Whatman
®
indicator papers,
pH Indicators, Colour Bonded, 0.0 to 14.0 range
Expand
View Pricing
WHA2614991
Whatman
®
indicator papers,
pH Indicators, Colour Bonded, 4.5 to 10.0 range
Expand
View Pricing
WHA2629990
Whatman
®
indicator papers,
pH Indicators, Integral Comparison Strip, 6.0 to 8.1 range
Expand
View Pricing
WHA2600100A
Whatman
®
indicator papers,
pH Indicators, Standard Full Range, 1.0 to 14.0 range
Expand
View Pricing
WHA2600102A
Whatman
®
indicator papers,
pH Indicators, Standard Narrow Range, 4.0 to 7.0 range
Expand
View Pricing
WHA2602501A
Whatman
®
indicator papers,
Specialized Test Papers, Lead Acetate
Expand
View Pricing
WHA2612990
Whatman
®
indicator papers,
pH Indicators, Integral Comparison Strip, 1.0 to 12.0 range
Expand
View Pricing
WHA2600104A
Whatman
®
indicator papers,
pH Indicators, Standard Narrow Range, 8.0 to 10.0 range
Expand
View Pricing
WHA2602500A
Whatman
®
indicator papers,
Specialized Test Papers, Potassium Iodide
Expand
View Pricing
WHA10362010
Whatman
®
indicator papers,
pH range 2-9, non-bleeding
Expand
View Pricing
WHA10362030
Whatman
®
indicator papers,
pH range 0-11
Expand
View Pricing
WHA2626990
Whatman
®
indicator papers,
pH Indicators, Integral Comparison Strip, 1.8 to 3.8 range
Expand
View Pricing
WHA2651500
Whatman
®
indicator papers,
Starch iodide test paper book, book of 20 strips
Expand
View Pricing
WHA2600101A
Whatman
®
indicator papers,
pH Indicators, Standard Narrow Range, 0.5 to 5.5 range
Expand
View Pricing
WHA10360300
Whatman
®
indicator papers,
Indicator paper, Litmus blue pH 8-5
Expand
View Pricing
WHA2630990
Whatman
®
indicator papers,
pH Indicators, Integral Comparison Strip, 8.0 to 9.7 range
Expand
View Pricing
WHA2600103A
Whatman
®
indicator papers,
pH Indicators, Standard Narrow Range, 6.4 to 8.0 range
Expand
View Pricing
WHA2631990
Whatman
®
indicator papers,
pH indicators, integral comparison strip, 9.5 to 12.0 range
Expand
View Pricing
WHA2600204A
Whatman
®
indicator papers,
Acid Alkali Test Papers, Phenophthalein
Expand
View Pricing
Page 1 of 2
Page 1 of 2