Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Water-
18
O
Linear Formula:
H
2
18
O
CAS Number:
14314-42-2
Molecular Weight:
20.02
Products
Product Category
stable isotopes (7)
Brand
Sigma-Aldrich (6)
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Isotope
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
329878
Water-
18
O,
(for PET), 97 atom %
18
O
Expand
View Pricing
487090
Water-
18
O,
99 atom %
18
O
Expand
View Pricing
902187
Water-
18
O,
(for PET), ≥98 atom %
18
O
Expand
View Pricing
332089
Water-
18
O,
10 atom %
18
O
Expand
View Pricing
603112
Water-
18
O,
98 atom %
18
O
Expand
View Pricing
603147
Water-
18
O,
80 atom %
18
O
Expand
View Pricing
603139
Water-
18
O,
70 atom %
18
O
Expand
View Pricing