Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vornado
™
Vortexer
Products
Color
Manufacturer
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z741409
Vornado
™
Vortexer,
red, AC/DC input 230 V AC, EuroPlug
Expand
View Pricing
Z741408
Vornado
™
Vortexer,
purple, AC/DC input 230 V AC, EuroPlug
Expand
View Pricing
Z741413
Vornado
™
Vortexer,
purple, AC/DC input 230 V AC, UK plug
Expand
View Pricing
Z741407
Vornado
™
Vortexer,
green, AC/DC input 230 V AC, EuroPlug
Expand
View Pricing
Z741411
Vornado
™
Vortexer,
blue, AC/DC input 230 V AC, UK plug
Expand
View Pricing
Z741406
Vornado
™
Vortexer,
blue, AC/DC input 230 V AC, EuroPlug
Expand
View Pricing
Z741405
Vornado
™
Vortexer,
gray, AC/DC input 230 V AC, EuroPlug
Expand
View Pricing
Z741412
Vornado
™
Vortexer,
green, AC/DC input 230 V AC, UK plug
Expand
View Pricing
Z741414
Vornado
™
Vortexer,
red, AC/DC input 230 V AC, UK plug
Expand
View Pricing
Z741410
Vornado
™
Vortexer,
gray, AC/DC input 230 V AC, UK plug
Expand
View Pricing