Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Urea-
13
C,
15
N
2
solution
Empirical Formula (Hill Notation):
13
CH
4
15
N
2
O
Molecular Weight:
63.03
Products
Brand
Supelco (2)
Formula Weight
Format
Quality Segment
Isotope
Special Grade
analytical standard (2)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
611387
Urea-
13
C,
15
N
2
solution,
99 atom %
13
C, 98 atom %
15
N, 0.1 M in DMSO-
d
6
(99.9 atom % D), NMR tube size 5 mm × 7 in.
Expand
View Pricing
611360
Urea-
13
C,
15
N
2
solution,
0.1 M in DMSO-
d
6
(99.9 atom % D), NMR tube size 8 mm × 7 in.
Expand
View Pricing