
Tryptic Soy Broth Ready-to-use Bottles

Products
Product Category
Brand
Physical Form
Sterilization Compatibility
Agency
Chemical Composition
Capacity (mL)
Compatibility
Quality Segment
Closure Type
Application
Package Size
Sterility
Sterilization Method
Showing 1-20 of 24
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2