
Trandolapril Related Compound E

Synonym(s):
(2S,3aR,7aS)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]-1-oxopropyl]octahydro 1H-indole-2-carboxylic acid, Trandolapril diacid, Trandolaprilat
Empirical Formula (Hill Notation):
C22H30N2O5
CAS Number:
Molecular Weight:
402.48
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing