Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TOYOPEARL
®
HW-55F Bulk Media
Synonym(s):
TOYOPEARL
®
HW-55 Size Exclusion Media
Products
Product Category
resins (1)
Brand
Supelco (1)
Manufacturer
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
807457
TOYOPEARL
®
HW-55F Bulk Media,
phase hydroxylated methacrylate, bottle of 500 mL, 45 μm particle size
Expand
View Pricing
819812
TOYOPEARL
®
HW-55F Bulk Media,
phase hydroxylated methacrylate, bottle of 150 mL, 45 μm particle size
Expand
View Pricing
814687
TOYOPEARL
®
HW-55F Bulk Media,
phase hydroxylated methacrylate, bottle of 1 L, 45 μm particle size
Expand
View Pricing