Skip to Content
Merck
Tetraoctylammonium tetrafluoroborate ≥97.0%

Tetraoctylammonium tetrafluoroborate

Tetraoctylammonium tetrafluoroborate ≥97.0%
Linear Formula:
[CH3(CH2)7]4N(BF4)
CAS Number:
Molecular Weight:
553.69

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing