Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
tert-Octylamine
Synonym(s):
tert-Octylamine, (2,4,4-trimethylpentan-2-amine), 1,1,3,3-Tetramethylbutylamine, 2-Amino-2,4,4-trimethylpentane
Linear Formula:
(CH
3
)
3
CCH
2
C(CH
3
)
2
NH
2
CAS Number:
107-45-9
Molecular Weight:
129.24
EC Number:
203-491-1
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Purity
Agency
Format
Application
Markush Class
Markush Group
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1A01310
tert-Octylamine,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
O6000
tert-Octylamine,
95%
Expand
View Pricing