
tert-Butyl acetate

Synonym(s):
TBAc, tert-Butyl acetate
Linear Formula:
CH3COOC(CH3)3
CAS Number:
Molecular Weight:
116.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing