
Tectorigenin

Synonym(s):
4′,5,7-Trihydroxy 6-methoxyisoflavone, 5,7-Dihydroxy-3-(4-hydroxy-phenyl)-6-methoxy-chromen-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, 5-Hydroxyglycitein, Tectrigenin
Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing