Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tecan
®
-style liquid level sensing tips
Products
Product Category
pipette tips (9)
Manufacturer
Color
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
AXYTT50CBKHTRS
Tecan
®
-style liquid level sensing tips,
size 50 μL, pkg of 24x96tips/cs, racked, sterile
Expand
View Pricing
AXYTT50CBKHTR36
Tecan
®
-style liquid level sensing tips,
size 50 μL, case of 6912 ea 36x192tips/cs, 2 racks in cardboard box
Expand
View Pricing
AXYTT50CBKHTR
Tecan
®
-style liquid level sensing tips,
size 50 μL, pkg of 24x96tips/cs, rack in blister box
Expand
View Pricing
AXYTT200CBKHTRS
Tecan
®
-style liquid level sensing tips,
size 200 μL, case of 2304 ea 24x96tips/cs, racked, sterile
Expand
View Pricing
AXYTT1000CBKHTR
Tecan
®
-style liquid level sensing tips,
size 1000 μL, pkg of 16x96tips/cs, rack in blister box
Expand
View Pricing
AXYTT200CBKHTR
Tecan
®
-style liquid level sensing tips,
size 200 μL, pkg of 24x96tips/cs, rack in blister pack
Expand
View Pricing
AXYTT20CBKHTR
Tecan
®
-style liquid level sensing tips,
size 20 μL, pkg of 24x96tips/cs, rack in blister box
Expand
View Pricing
AXYTT1000CBKHTRS
Tecan
®
-style liquid level sensing tips,
size 1000 μL, pkg of 16x96tips/cs, racked, sterile
Expand
View Pricing
AXYTT1000CBKHTR24
Tecan
®
-style liquid level sensing tips,
size 1000 μL, pkg of 24x192tips/cs, 2 racks in cardboard box
Expand
View Pricing