Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 96
Page 1 of 5
Page 1 of 5
Compare
Product Number
Description
Pricing
GF00247575
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
rod, 100mm x 9mm diameter, as drawn
Expand
View Pricing
GF38541449
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
rod, 500mm x 2.0mm diameter, annealed
Expand
View Pricing
GF41015113
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
foil, 0.1m coil, 100mm coil width, 0.50mm thickness, annealed
Expand
View Pricing
GF14269811
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
rod, 100mm x 6mm diameter, as drawn
Expand
View Pricing
GF26456441
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
rod, 200mm x 2.0mm diameter, annealed
Expand
View Pricing
GF75000349
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
rod, 100mm x 12mm diameter, as drawn
Expand
View Pricing
GF29844872
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
foil, 0.1m coil, 100mm coil width, 0.25mm thickness, annealed
Expand
View Pricing
GF94020319
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
rod, 100mm x 2.0mm diameter, annealed
Expand
View Pricing
GF14346165
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
100m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF53819297
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
rod, 500mm x 12mm diameter, as drawn
Expand
View Pricing
GF08733955
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
rod, 200mm x 9mm diameter, as drawn
Expand
View Pricing
GF83726893
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
rod, 200mm x 6mm diameter, as drawn
Expand
View Pricing
GF21904208
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
insulated wire, 20m length, 0.05mm conductor diameter, 0.002mm insulation thickness, polyester insulation
Expand
View Pricing
GF83169322
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
rod, 1000mm x 2.0mm diameter, annealed
Expand
View Pricing
GF51869122
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
insulated wire, 20m length, 0.125mm conductor diameter, 0.008mm insulation thickness, polyimide insulation
Expand
View Pricing
GF19212836
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
50m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF72153705
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
10m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF60480581
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
2000m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF55395872
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
500m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF38924387
T2 - thermocouple alloy, Ni95(Al+Mn+Si)5,
rod, 200mm x 12mm diameter, as drawn
Expand
View Pricing
Page 1 of 5
Page 1 of 5