Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Supelclean
™
ENVI-WAX SPE Tube
Synonym(s):
Supelclean
™
ENVI-WAX SPE
Products
Brand
Supelco (2)
Agency
Chemical Composition
Separation Technique
Volume (ml)
Quality Segment
Application
Package Size
Particle Size (µm)
Pore Size (Å)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
54057-U
Supelclean
™
ENVI-WAX SPE Tube,
suitable for PFAS testing, bed wt. 500 mg, volume 6 mL, pk of 30 ea
Expand
View Pricing
54056-U
Supelclean
™
ENVI-WAX SPE Tube,
bed wt. 200 mg, suitable for PFAS testing, volume 6 mL, pk of 30 ea
Expand
View Pricing