Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sulfonyl chloride, polymer-bound
CAS Number:
163894-16-4
Products
Brand
Sigma-Aldrich (2)
Quality Segment
Chain Type
Reaction Type
Reactivity
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
516228
Sulfonyl chloride, polymer-bound,
70-90 mesh, extent of labeling: 2.5-3.0 mmol/g loading, 8.5 % cross-linked with divinylbenzene
Expand
View Pricing
498211
Sulfonyl chloride, polymer-bound,
100-200 mesh, extent of labeling: 1.5-2.0 mmol/g loading, 1 % cross-linked
Expand
View Pricing