
Sulfadiazine for identification of impurity F

Synonym(s):
Sulfadiazine, 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide, N1-(Pyrimidin-2-yl)sulfanilamide
Empirical Formula (Hill Notation):
C10H10N4O2S
CAS Number:
Molecular Weight:
250.28
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing