
Sulbactam Related Compound F

Synonym(s):
(2S,5R)-6,6-Dibromo-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6,6-Dibromopenicillanic acid
Empirical Formula (Hill Notation):
C8H9Br2NO3S
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing