Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Suba-Seal
®
septa
Products
Product Category
septa (42)
Brand
Sigma-Aldrich (37)
Color
Filter Products
Showing 1-20 of 42
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
Z167282
Suba-Seal
®
septa,
white rubber, Suba-Seal
®
25, fits neck I.D., 14 mm, for use with 14/20 joints
Expand
View Pricing
Z124591
Suba-Seal
®
septa,
red rubber, Suba-Seal
®
25, fits neck I.D., 14 mm, for use with 14/20 joints
Expand
View Pricing
Z167320
Suba-Seal
®
septa,
white rubber, Suba-Seal
®
45, fits neck I.D., 22 mm, for use with 24/40 joints
Expand
View Pricing
Z124656
Suba-Seal
®
septa,
red rubber, Suba-Seal
®
45, fits neck I.D., 22 mm, for use with 24/40 joints
Expand
View Pricing
Z167304
Suba-Seal
®
septa,
white rubber, Suba-Seal
®
37, fits neck I.D., 19 mm, for use with 19/22 joints
Expand
View Pricing
Z124621
Suba-Seal
®
septa,
red rubber, Suba-Seal
®
37, fits neck I.D., 19 mm, for use with 19/22 joints
Expand
View Pricing
Z167355
Suba-Seal
®
septa,
white rubber, Suba-Seal
®
57, fits neck I.D., 27 mm, for use with 29/42 joints
Expand
View Pricing
Z124664
Suba-Seal
®
septa,
red rubber, Suba-Seal
®
49, fits neck I.D., 24 mm
Expand
View Pricing
Z124567
Suba-Seal
®
septa,
red rubber, Suba-Seal
®
13, fits neck I.D., 9.5 mm, for use with 10/30 joints
Expand
View Pricing
Z124680
Suba-Seal
®
septa,
red rubber, Suba-Seal
®
57, fits neck I.D., 27 mm, for use with 29/42 joints
Expand
View Pricing
Z124575
Suba-Seal
®
septa,
red rubber, Suba-Seal
®
17, fits neck I.D., 11 mm
Expand
View Pricing
Z167258
Suba-Seal
®
septa,
white rubber, Suba-Seal
®
13, fits neck I.D., 9.5 mm, for use with 10/30 joints
Expand
View Pricing
Z558192
Suba-Seal
®
septa,
red rubber, Suba-Seal
®
61, fits neck I.D., 29 mm
Expand
View Pricing
Z558311
Suba-Seal
®
septa,
blue rubber, Suba-Seal
®
57, fits neck I.D., 27 mm, for use with 29/42 joints
Expand
View Pricing
Z167347
Suba-Seal
®
septa,
white rubber, Suba-Seal
®
53, fits neck I.D., 25.5 mm
Expand
View Pricing
Z648906
Suba-Seal
®
septa,
blue rubber, Suba-Seal
®
25, fits neck I.D., 14 mm, for use with 14/20 joints
Expand
View Pricing
Z124559
Suba-Seal
®
septa,
red rubber, Suba-Seal
®
9, fits neck I.D., 8 mm
Expand
View Pricing
Z167231
Suba-Seal
®
septa,
white rubber, Suba-Seal
®
9, fits neck I.D., 8 mm
Expand
View Pricing
Z124605
Suba-Seal
®
septa,
red rubber, Suba-Seal
®
29, fits neck I.D., 16 mm
Expand
View Pricing
Z124613
Suba-Seal
®
septa,
red rubber, Suba-Seal
®
33, fits neck I.D., 17.5 mm, for use with all Sure/Seal bottles
Expand
View Pricing
Page 1 of 3
Page 1 of 3