Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SPE Tube Adapter
Products
Product Category
adapters (4)
Brand
Supelco (3)
Application
Package Size
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
57020-U
SPE Tube Adapter,
for use with 1, 3, & 6 mL tubes, pk of 12
Expand
View Pricing
57126
SPE Tube Adapter,
configured for AutoTrace Automated Systems, for use with 6mL tubes, pk of 6
Expand
View Pricing
504335
SPE Tube Adapter,
PTFE (with female luer port), for use with 6 mL glass SPE tubes, pk of 24
Expand
View Pricing
57123
SPE Tube Adapter,
configured for AutoTrace Automated Systems, for use with 3mL tubes, pk of 6
Expand
View Pricing