Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
Sterileware
™
Volumetric Sampling Spoons
Products
Product Category
spatulas/scoops (4)
Manufacturer
Capacity (mL)
Diameter (mm)
Length (cm)
Length (in)
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
BAF369430010
SP Bel-Art
®
Sterileware
™
Volumetric Sampling Spoons,
white high-density polyethylene, capacity 10 mL (0.34 oz), L 17.1 cm (6.75 in.), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369430020
SP Bel-Art
®
Sterileware
™
Volumetric Sampling Spoons,
white high-density polyethylene, capacity × L 20 mL (0.68 oz) × 18 cm (7.125 in.), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369430001
SP Bel-Art
®
Sterileware
™
Volumetric Sampling Spoons,
white high-density polyethylene, capacity 1 mL (0.03 oz), L 16.5 cm (6.5 in.), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369430005
SP Bel-Art
®
Sterileware
™
Volumetric Sampling Spoons,
white high-density polyethylene, capacity 5 mL (0.17 oz), L 16.5 cm (6.5 in.), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing