Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
Sterileware
™
Flat Bottom Sampling Scoop
Products
Product Category
spatulas/scoops (7)
Manufacturer
Height (mm)
Length (in)
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
BAF369110025
SP Bel-Art
®
Sterileware
™
Flat Bottom Sampling Scoop,
white polystyrene, nominal capacity 25 mL (0.8 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369111000
SP Bel-Art
®
Sterileware
™
Flat Bottom Sampling Scoop,
white polystyrene, nominal capacity 1000 mL (34 oz), sterile, pack of 30 ea (individually wrapped)
Expand
View Pricing
BAF369110050
SP Bel-Art
®
Sterileware
™
Flat Bottom Sampling Scoop,
white polystyrene, nominal capacity 50 mL (1.7 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369110500
SP Bel-Art
®
Sterileware
™
Flat Bottom Sampling Scoop,
white polystyrene, nominal capacity 500 mL, sterile, pack of 40 ea (individually wrapped)
Expand
View Pricing
BAF369110150
SP Bel-Art
®
Sterileware
™
Flat Bottom Sampling Scoop,
white polystyrene, nominal capacity 150 mL (5.1 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369110100
SP Bel-Art
®
Sterileware
™
Flat Bottom Sampling Scoop,
white polystyrene, nominal capacity 100 mL (3.4 oz), sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing
BAF369110250
SP Bel-Art
®
Sterileware
™
Flat Bottom Sampling Scoop,
white polystyrene, nominal capacity 250 mL, sterile, pack of 100 ea (individually wrapped)
Expand
View Pricing