Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Span
®
80
Synonym(s):
Span
®
80, Sorbitan monooleate
Empirical Formula (Hill Notation):
C
24
H
44
O
6
CAS Number:
1338-43-8
Molecular Weight:
428.60
EC Number:
215-665-4
Products
Product Category
detergents (2)
Brand
Sigma-Aldrich (1)
Supelco (2)
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
8.40123
Span
®
80,
for synthesis
Expand
View Pricing
85548
Span
®
80,
viscosity 1000-2000 mPa.s (20 °C)
Expand
View Pricing
09569
Span
®
80,
suitable for GC
Expand
View Pricing